Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACT12
Entrez Gene ID
823805
Gene Name
actin-12
Gene Aliases
ACTIN, AT3G46520, actin-12
Chromosome Location
Chr.3: 17128211 - 17131466 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339262.1 NP_001327017.1 3 - 4 595 128
NM_114519.3 NP_190236.1 3 - 4 595 128
GenBank mRNA BT003847.1 - 3 - 4 543 128
BT005073.1 - 2 - 3 453 128

View Full Product Details