Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA4
Entrez Gene ID
510423
Gene Name
proteasome 20S subunit alpha 4
Gene Aliases
-
Chromosome Location
Chr.21: 30920003 - 30927853 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034381.2 NP_001029553.1 6 - 7 457 81
GenBank mRNA BC102102.1 - 6 - 7 459 81

View Full Product Details