Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GM2A
Entrez Gene ID
504524
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.7: 62495947 - 62508685 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205547.3 NP_001192476.2 2 - 3 385 63

View Full Product Details