Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hlh-2
Entrez Gene ID
172458
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_M05B5.5
Chromosome Location
Chr.I: 7190788 - 7193924 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026410.4 NP_001021581.1 2 - 3 654 91
NR_002347.1 3 - 4 593 91

View Full Product Details