Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcp-1
Entrez Gene ID
179049
Gene Name
HoloCentric chromosome binding Protein
Gene Aliases
CELE_ZK1055.1
Chromosome Location
Chr.V: 6594194 - 6599208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072276.4 NP_504677.1 7 - 8 4234 66

View Full Product Details