Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-48
Entrez Gene ID
184964
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F26D2.9
Chromosome Location
Chr.V: 16420996 - 16422774 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074749.2 NP_507150.2 3 - 4 481 69

View Full Product Details