The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243989.1 | NP_001230918.1 | 20 - 21 | 2683 | 79 |
XM_007646774.1 | XP_007644964.1 | 21 - 22 | 2847 | 79 | |
XM_007646775.1 | XP_007644965.1 | 19 - 20 | 2622 | 79 | |
GenBank mRNA | M17896.1 | - | 7 - 8 | 812 | 79 |
M60041.1 | - | 20 - 21 | 2683 | 79 |