Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pidd
Entrez Gene ID
100753147
Gene Name
p53-induced death domain protein
Gene Aliases
Lrdd
Chromosome Location
Chr.Un|NW_003614763: 285519 - 291344 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509765.2 XP_003509813.1 14 - 15 2510 89
XM_007650419.1 XP_007648609.1 14 - 15 2415 89
XM_007650420.1 XP_007648610.1 14 - 15 2506 89
XM_007650421.1 XP_007648611.1 14 - 15 2554 89

View Full Product Details