Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpc7
Entrez Gene ID
100755860
Gene Name
transient receptor potential cation channel, subfamily C, member 7
Gene Aliases
I79_010635
Chromosome Location
Chr.Un|NW_003613997: 521431 - 675280 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644313.1 XP_007642503.1 12 - 13 2773 67

View Full Product Details