Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PATE2
Entrez Gene ID
102183939
Gene Name
prostate and testis expressed 2
Gene Aliases
-
Chromosome Location
Chr.29: 28541239 - 28542296 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699716.1 XP_005699773.1 2 - 3 141 78

View Full Product Details