Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tango11
Entrez Gene ID
246596
Gene Name
Transport and Golgi organization 11
Gene Aliases
BEST:LD33633, BcDNA:LD22567, CG30404, Dmel\CG30404, Dmel_CG30404, TANGO11, anon-EST:fe1F11, tango11
Chromosome Location
Chr.2R: 21716780 - 21719963 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050404
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_166481.3 NP_726111.1 2 - 3 708 91
NM_166482.2 NP_726112.1 1 - 2 761 91
GenBank mRNA AY051672.1 - 2 - 3 676 91

View Full Product Details