Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tutl
Entrez Gene ID
46015
Gene Name
turtle
Gene Aliases
CG15426, CG15427, CG15428, CT35489, CT35490, Dmel\CG15427, Dmel_CG15427, Tutl, jf4, l(2)01085, l(2)24Df, l(2)jf4, mdcds_7441
Chromosome Location
Chr.2L: 4283148 - 4323781 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010473
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273090.1 NP_001260019.1 6 - 7 1587 78
NM_164578.4 NP_722969.1 6 - 7 1529 78
GenBank mRNA AJ312132.1 - 6 - 7 1484 78

View Full Product Details