Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETHR
Entrez Gene ID
42523
Gene Name
CG5911-PA
Gene Aliases
CG5911, CG5911a, CG5911b, Dm ETHRa, Dmel\CG5911, Dmel_CG5911, anon-WO0170980.37, anon-WO0170980.38
Chromosome Location
Chr.3R: 21247011 - 21265134 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038874
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300510.1 NP_001287439.1 2 - 3 1263 88
NM_142703.3 NP_650960.2 2 - 3 1256 88
NM_206533.2 NP_996255.1 2 - 3 1256 88
GenBank mRNA AY220741.1 - 1 - 2 89 88
AY220742.1 - 1 - 2 89 88

View Full Product Details