Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bili
Entrez Gene ID
43019
Gene Name
Band4.1 inhibitor LRP interactor
Gene Aliases
CG11848, Dmel\CG11848, Dmel_CG11848, dBili
Chromosome Location
Chr.3R: 25131306 - 25136709 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170251.2 NP_001163722.1 6 - 7 1844 93
NM_001170252.2 NP_001163723.1 6 - 7 1833 93
NM_001170253.2 NP_001163724.1 6 - 7 1928 93
NM_001170254.2 NP_001163725.1 6 - 7 1864 93
NM_001170255.1 NP_001163726.1 6 - 7 1984 93
NM_001300592.1 NP_001287521.1 6 - 7 2237 93
NM_143085.3 NP_651342.3 6 - 7 1941 93
GenBank mRNA AY094737.1 - 6 - 7 1903 93

View Full Product Details