Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sncgb
Entrez Gene ID
553679
Gene Name
synuclein, gamma b (breast cancer-specific protein 1)
Gene Aliases
sncg, zgc:112131
Chromosome Location
Chr.12: 25893006 - 25916598 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020652.1 NP_001018488.1 2 - 3 309 75
XM_021480112.1 XP_021335787.1 2 - 3 375 75
GenBank mRNA BC095672.1 - 2 - 3 309 75
EU368946.1 - 2 - 3 381 75

View Full Product Details