Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnajc9
Entrez Gene ID
436706
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 9
Gene Aliases
zgc:92648
Chromosome Location
Chr.12: 233882 - 243917 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002433.1 NP_001002433.1 4 - 5 724 61
GenBank mRNA BC076133.1 - 4 - 5 724 61

View Full Product Details