Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- adssl
- Entrez Gene ID
-
192305
- Gene Name
- adenylosuccinate synthase, like
- Gene Aliases
-
adss, adss2, chunp6912, zgc:66318
- Chromosome Location
-
Chr.21: 26522638 - 26531032 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY099534.1 |
- |
11 - 12 |
1202 |
66 |
View Full Product Details