Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY28
Entrez Gene ID
100170676
Gene Name
transcription factor
Gene Aliases
-
Chromosome Location
Chr.2: 11099620 - 11103046 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250598.1 NP_001237527.1 3 - 3 1052 96
GenBank mRNA EU375345.1 - 3 - 3 1052 96

View Full Product Details