Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278503.1 | NP_001265432.1 | 10 | 1117 | |
NM_001278504.1 | NP_001265433.1 | 8 | 906 | ||
NM_152713.4 | NP_689926.1 | 9 | 1029 | ||
XM_011542807.2 | XP_011541109.1 | 9 | 1056 | ||
GenBank mRNA | AK130548.1 | - | 2 | 142 | |
AK290040.1 | - | 9 | 979 | ||
AK290657.1 | - | 9 | 977 | ||
AK295892.1 | - | 8 | 834 | ||
AK308902.1 | - | 9 | 972 | ||
BC020965.2 | - | 9 | 962 | 3891543 | |
BC048348.1 | - | 8 | 915 | 5561615 | |
BT007100.1 | - | 8 | 870 | ||
BX649102.1 | - | 10 | 1055 | ||
L38961.1 | - | 9 | 976 | ||
L47337.1 | - | 9 | 986 |