Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278503.1 | NP_001265432.1 | 17 | 2152 | |
NM_001278504.1 | NP_001265433.1 | 15 | 1941 | ||
NM_152713.4 | NP_689926.1 | 16 | 2064 | ||
XM_011542807.2 | XP_011541109.1 | 16 | 2091 | ||
GenBank mRNA | AK290040.1 | - | 16 | 2014 | |
AK290657.1 | - | 16 | 2012 | ||
AK295892.1 | - | 15 | 1869 | ||
BC020965.2 | - | 16 | 1997 | 3891543 | |
BC048348.1 | - | 15 | 1950 | 5561615 | |
BT007100.1 | - | 15 | 1905 | ||
BX649102.1 | - | 17 | 2090 | ||
L38961.1 | - | 16 | 2011 | ||
L47337.1 | - | 16 | 2021 |