Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LYRM1
- Entrez Gene ID
-
57149
- Gene Name
- LYR motif containing 1
- Gene Aliases
-
A211C6.1
- Chromosome Location
-
Chr.16: 20899626 - 20925006 on Build GRCh38
View Full Product Details