Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21B
Entrez Gene ID
25895
Gene Name
methyltransferase like 21B
Gene Aliases
FAM119B
Chromosome Location
Chr.12: 57772600 - 57782541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK024983.1 - 2 - 3 301 84

View Full Product Details