The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253693.1 | NP_001240622.1 | 3 - 4 | 528 | 85 |
NM_018298.10 | NP_060768.8 | 4 - 5 | 696 | 85 | |
XM_005271003.1 | XP_005271060.1 | 5 - 6 | 1093 | 85 | |
XM_006710750.1 | XP_006710813.1 | 5 - 6 | 912 | 85 | |
XM_011541739.2 | XP_011540041.2 | 4 - 5 | 620 | 85 | |
XM_011541740.2 | XP_011540042.1 | 3 - 4 | 454 | 85 | |
XM_011541741.2 | XP_011540043.1 | 4 - 5 | 627 | 85 | |
XM_011541742.1 | XP_011540044.1 | 5 - 6 | 830 | 85 | |
GenBank mRNA | AF475085.1 | - | 4 - 5 | 602 | 85 |
AK001868.1 | - | 3 - 4 | 434 | 85 | |
AK292206.1 | - | 4 - 5 | 628 | 85 | |
AK314354.1 | - | 4 - 5 | 656 | 85 | |
BC060765.1 | - | 5 - 6 | 926 | 85 | |
BX647497.1 | - | 6 - 7 | 849 | 85 |