Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLYR1
Entrez Gene ID
84656
Gene Name
glyoxylate reductase 1 homolog
Gene Aliases
BM045, HIBDL, N-PAC, NP60
Chromosome Location
Chr.16: 4803203 - 4847388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308096.1 NP_001295025.1 3 - 4 195 80
NM_001324096.1 NP_001311025.1 3 - 4 281 80
NM_001324097.1 NP_001311026.1 3 - 4 281 80
NM_001324098.1 NP_001311027.1 3 - 4 281 80
NM_032569.3 NP_115958.2 3 - 4 195 80
NR_136696.1 3 - 4 281 80
NR_136697.1 3 - 4 281 80
NR_136698.1 3 - 4 281 80
NR_136699.1 3 - 4 281 80
NR_136700.1 3 - 4 281 80
XM_005255639.4 XP_005255696.2 3 - 4 239 80
XM_011522716.2 XP_011521018.1 3 - 4 323 80
XM_017023786.1 XP_016879275.1 3 - 4 276 80
XM_017023787.1 XP_016879276.1 3 - 4 276 80
GenBank mRNA AF244907.1 - 3 - 4 176 80
AF326966.1 - 3 - 4 158 80
AK296842.1 - 3 - 4 172 80
AK307834.1 - 3 - 4 171 80
AY352585.1 - 3 - 4 158 80
BC003693.1 - 2 - 3 120 80
BC047223.1 - 3 - 4 175 80
DB492328.1 - 3 - 4 200 80
HY130375.1 - 3 - 4 282 80

View Full Product Details