The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000875.4 | NP_000866.1 | 20 - 21 | 4760 | 99 |
NM_001291858.1 | NP_001278787.1 | 20 - 21 | 4757 | 99 | |
XM_011521516.2 | XP_011519818.1 | 19 - 20 | 3210 | 99 | |
XM_011521517.2 | XP_011519819.1 | 16 - 17 | 2623 | 99 | |
XM_017022136.1 | XP_016877625.1 | 20 - 21 | 4263 | 99 | |
XM_017022137.1 | XP_016877626.1 | 20 - 21 | 4263 | 99 | |
XM_017022138.1 | XP_016877627.1 | 20 - 21 | 4260 | 99 | |
XM_017022139.1 | XP_016877628.1 | 20 - 21 | 3701 | 99 | |
GenBank mRNA | AB425196.1 | - | 20 - 21 | 4243 | 99 |
BC113610.1 | - | 20 - 21 | 3749 | 99 | |
BC113612.1 | - | 20 - 21 | 3749 | 99 | |
BC143721.1 | - | 20 - 21 | 3746 | 99 | |
X04434.1 | - | 20 - 21 | 3762 | 99 |