The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006467.2 | NP_006458.2 | 1 - 1 | 94 | |
XM_005248404.3 | XP_005248461.1 | 1 - 1 | 94 | ||
XM_011543102.2 | XP_011541404.1 | 1 - 1 | 94 | ||
XM_011543103.2 | XP_011541405.1 | 1 - 1 | 94 | ||
XM_011543104.2 | XP_011541406.1 | 1 - 1 | 94 | ||
XM_017008957.1 | XP_016864446.1 | 1 - 1 | 94 | ||
GenBank mRNA | AK295434.1 | - | 1 - 1 | 53 | 94 |
BM788728.1 | - | 1 - 1 | 53 | 94 |