The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253866.1 | NP_001240795.1 | 10 - 11 | 1344 | 84 |
NM_014263.3 | NP_055078.1 | 11 - 12 | 1443 | 84 | |
NM_139312.2 | NP_647473.1 | 12 - 13 | 1614 | 84 | |
XM_011519300.2 | XP_011517602.1 | 11 - 12 | 1897 | 84 | |
GenBank mRNA | AF070656.1 | - | 7 - 8 | 758 | 84 |
AF151782.1 | - | 12 - 13 | 1458 | 84 | |
AJ132637.1 | - | 11 - 12 | 1416 | 84 | |
AK001259.1 | - | 2 - 3 | 205 | 84 | |
AK291292.1 | - | 11 - 12 | 1372 | 84 | |
AK297973.1 | - | 10 - 11 | 1308 | 84 | |
AK309708.1 | - | 7 - 8 | 889 | 84 | |
AK314372.1 | - | 11 - 12 | 1371 | 84 | |
AY358484.1 | - | 11 - 12 | 1369 | 84 | |
BC007795.2 | - | 12 - 13 | 1550 | 84 | |
BC023507.2 | - | 11 - 12 | 1385 | 84 | |
BC024032.2 | - | 11 - 12 | 1390 | 84 |