The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253866.1 | NP_001240795.1 | 7 - 8 | 970 | 194 |
NM_014263.3 | NP_055078.1 | 8 - 9 | 1069 | 194 | |
NM_139312.2 | NP_647473.1 | 9 - 10 | 1240 | 194 | |
XM_011519300.2 | XP_011517602.1 | 8 - 9 | 1523 | 194 | |
GenBank mRNA | AF070656.1 | - | 4 - 5 | 384 | 194 |
AF151782.1 | - | 9 - 10 | 1084 | 194 | |
AJ132637.1 | - | 8 - 9 | 1042 | 194 | |
AK291292.1 | - | 8 - 9 | 998 | 194 | |
AK297973.1 | - | 7 - 8 | 934 | 194 | |
AK309708.1 | - | 4 - 5 | 516 | 193 | |
AK314372.1 | - | 8 - 9 | 997 | 194 | |
AY358484.1 | - | 8 - 9 | 995 | 194 | |
BC007795.2 | - | 9 - 10 | 1176 | 194 | |
BC023507.2 | - | 8 - 9 | 1011 | 194 | |
BC024032.2 | - | 8 - 9 | 1016 | 194 |