Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPM2AIP1
Entrez Gene ID
9852
Gene Name
EPM2A interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 36985866 - 36993304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014805.3 NP_055620.1 1 - 1 5025 114
GenBank mRNA AY178832.1 - 1 - 1 5025 114
BC041957.1 - 1 - 1 797 114
BC064696.1 - 1 - 1 4892 114

View Full Product Details