Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21B
Entrez Gene ID
25895
Gene Name
methyltransferase like 21B
Gene Aliases
FAM119B
Chromosome Location
Chr.12: 57772600 - 57782541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206914.1 NP_996797.1 3 - 4 64
GenBank mRNA AK024983.1 - 3 - 4 437 63
AL050100.1 - 3 - 4 64

View Full Product Details