The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002158.3 | NP_002149.2 | 4 - 5 | 951 | 95 |
XM_005264282.2 | XP_005264339.1 | 3 - 4 | 825 | 95 | |
XM_005264283.2 | XP_005264340.1 | 5 - 6 | 1146 | 95 | |
XM_006712001.3 | XP_006712064.1 | 4 - 5 | 882 | 95 | |
XM_006712002.3 | XP_006712065.1 | 3 - 4 | 842 | 95 | |
XM_011532808.2 | XP_011531110.1 | 5 - 6 | 1068 | 95 | |
XM_017003956.1 | XP_016859445.1 | 4 - 5 | 1286 | 95 | |
XM_017003957.1 | XP_016859446.1 | 5 - 6 | 1143 | 95 | |
XM_017003958.1 | XP_016859447.1 | 5 - 6 | 1142 | 95 | |
XM_017003959.1 | XP_016859448.1 | 5 - 6 | 989 | 95 | |
GenBank mRNA | BC063305.1 | - | 4 - 5 | 876 | 95 |
BC069731.1 | - | 2 - 3 | 444 | 95 | |
U57029.1 | - | 2 - 3 | 647 | 95 |