Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLYR1
Entrez Gene ID
84656
Gene Name
glyoxylate reductase 1 homolog
Gene Aliases
BM045, HIBDL, N-PAC, NP60
Chromosome Location
Chr.16: 4803203 - 4847388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308096.1 NP_001295025.1 1 - 2 74 64
NM_001324096.1 NP_001311025.1 1 - 2 160 64
NM_001324097.1 NP_001311026.1 1 - 2 160 64
NM_001324098.1 NP_001311027.1 1 - 2 160 64
NM_032569.3 NP_115958.2 1 - 2 74 64
NR_136695.1 1 - 2 160 64
NR_136696.1 1 - 2 160 64
NR_136697.1 1 - 2 160 64
NR_136698.1 1 - 2 160 64
NR_136699.1 1 - 2 160 64
NR_136700.1 1 - 2 160 64
XM_005255639.4 XP_005255696.2 1 - 2 118 64
XM_011522717.1 XP_011521019.1 1 - 2 60 64
XM_017023786.1 XP_016879275.1 1 - 2 155 64
XM_017023787.1 XP_016879276.1 1 - 2 155 64
XM_017023788.1 XP_016879277.1 1 - 2 60 64
XM_017023789.1 XP_016879278.1 1 - 2 60 64
GenBank mRNA AF244907.1 - 1 - 2 55 64
AK296842.1 - 1 - 2 51 64
AK307834.1 - 1 - 2 50 64
BC032855.1 - 1 - 2 65 64
BC047223.1 - 1 - 2 54 64
BC064940.1 - 1 - 2 59 64
HY130375.1 - 1 - 2 161 64

View Full Product Details