The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308096.1 | NP_001295025.1 | 2 - 3 | 105 | 67 |
NM_001324096.1 | NP_001311025.1 | 2 - 3 | 191 | 67 | |
NM_001324097.1 | NP_001311026.1 | 2 - 3 | 191 | 67 | |
NM_001324098.1 | NP_001311027.1 | 2 - 3 | 191 | 67 | |
NM_032569.3 | NP_115958.2 | 2 - 3 | 105 | 67 | |
NR_136695.1 | 2 - 3 | 191 | 67 | ||
NR_136696.1 | 2 - 3 | 191 | 67 | ||
NR_136697.1 | 2 - 3 | 191 | 67 | ||
NR_136698.1 | 2 - 3 | 191 | 67 | ||
NR_136699.1 | 2 - 3 | 191 | 67 | ||
NR_136700.1 | 2 - 3 | 191 | 67 | ||
XM_005255639.4 | XP_005255696.2 | 2 - 3 | 149 | 67 | |
XM_011522716.2 | XP_011521018.1 | 2 - 3 | 233 | 67 | |
XM_011522717.1 | XP_011521019.1 | 2 - 3 | 91 | 67 | |
XM_017023786.1 | XP_016879275.1 | 2 - 3 | 186 | 67 | |
XM_017023787.1 | XP_016879276.1 | 2 - 3 | 186 | 67 | |
XM_017023788.1 | XP_016879277.1 | 2 - 3 | 91 | 67 | |
XM_017023789.1 | XP_016879278.1 | 2 - 3 | 91 | 67 | |
GenBank mRNA | AF244907.1 | - | 2 - 3 | 86 | 67 |
AF326966.1 | - | 2 - 3 | 68 | 67 | |
AK296842.1 | - | 2 - 3 | 82 | 67 | |
AK307834.1 | - | 2 - 3 | 81 | 67 | |
AY352585.1 | - | 2 - 3 | 68 | 67 | |
BC003693.1 | - | 1 - 2 | 30 | 67 | |
BC032855.1 | - | 2 - 3 | 96 | 67 | |
BC047223.1 | - | 2 - 3 | 85 | 67 | |
BC064940.1 | - | 2 - 3 | 90 | 67 | |
HY130375.1 | - | 2 - 3 | 192 | 67 |