The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_136695.1 | 3 - 4 | 275 | 87 | |
XM_011522717.1 | XP_011521019.1 | 3 - 4 | 175 | 87 | |
XM_017023788.1 | XP_016879277.1 | 3 - 4 | 175 | 87 | |
XM_017023789.1 | XP_016879278.1 | 3 - 4 | 175 | 87 | |
GenBank mRNA | BC064940.1 | - | 3 - 4 | 174 | 87 |