Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
Chromosome Location
Chr.12: 56080025 - 56103507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001982.3 NP_001973.2 15 - 16 2135 80
GenBank mRNA AK291681.1 - 15 - 16 1988 80
AK294719.1 - 15 - 16 1992 80
AK295650.1 - 16 - 17 1902 80
AK300909.1 - 2 - 3 234 80
BC041579.1 - 3 - 4 311 80
BC082992.1 - 15 - 16 1974 80
M29366.1 - 15 - 16 1958 80
M34309.1 - 15 - 16 2057 80

View Full Product Details