The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001982.3 | NP_001973.2 | 17 - 18 | 2323 | 96 |
GenBank mRNA | AK294719.1 | - | 17 - 18 | 2180 | 96 |
AK295650.1 | - | 18 - 19 | 2090 | 96 | |
AK300909.1 | - | 4 - 5 | 422 | 96 | |
BC082992.1 | - | 17 - 18 | 2162 | 96 | |
M29366.1 | - | 17 - 18 | 2146 | 96 | |
M34309.1 | - | 17 - 18 | 2245 | 96 |