The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001982.3 | NP_001973.2 | 9 - 10 | 1385 | 63 |
GenBank mRNA | AK291681.1 | - | 9 - 10 | 1238 | 63 |
AK294719.1 | - | 9 - 10 | 1242 | 63 | |
AK295650.1 | - | 10 - 11 | 1152 | 63 | |
BC082992.1 | - | 9 - 10 | 1224 | 63 | |
M29366.1 | - | 9 - 10 | 1208 | 63 | |
M34309.1 | - | 9 - 10 | 1307 | 63 |