Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC22
Entrez Gene ID
55001
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.1: 54779696 - 54802379 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114108.1 NP_001107580.1 2 - 3 728 61
NM_017904.3 NP_060374.2 2 - 3 728 61
XM_011541671.2 XP_011539973.1 2 - 3 1839 61
XM_017001582.1 XP_016857071.1 2 - 3 1862 61
GenBank mRNA AK000626.1 - 2 - 3 658 61

View Full Product Details