Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYRM1
Entrez Gene ID
57149
Gene Name
LYR motif containing 1
Gene Aliases
A211C6.1
Chromosome Location
Chr.16: 20899626 - 20925006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020424.4 NP_065157.1 1 - 2 133 113
XM_011545902.1 XP_011544204.1 1 - 2 119 113
GenBank mRNA DA572182.1 - 1 - 2 99 113

View Full Product Details