The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019043.3 | NP_061916.3 | 7 - 8 | 1012 | 132 |
XM_006717451.2 | XP_006717514.1 | 6 - 7 | 710 | 132 | |
XM_011519514.2 | XP_011517816.1 | 7 - 8 | 1025 | 132 | |
GenBank mRNA | AB085852.1 | - | 5 - 6 | 691 | 132 |
AK000812.1 | - | 7 - 8 | 971 | 132 | |
AK303597.1 | - | 7 - 8 | 1572 | 132 | |
AK308916.1 | - | 7 - 8 | 993 | 132 | |
AY152730.1 | - | 6 - 7 | 778 | 132 | |
BC054516.1 | - | 6 - 7 | 790 | 132 |