Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUGP2
Entrez Gene ID
10147
Gene Name
SURP and G-patch domain containing 2
Gene Aliases
SFRS14
Chromosome Location
Chr.19: 18990887 - 19034052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017392.4 NP_001017392.2 5 - 6 2478 85
NM_001321697.1 NP_001308626.1 5 - 6 2634 85
NM_001321698.1 NP_001308627.1 5 - 6 2426 85
NM_001321699.1 NP_001308628.1 5 - 6 2426 85
NM_014884.4 NP_055699.2 5 - 6 2478 85
XM_017026137.1 XP_016881626.1 5 - 6 2634 85
GenBank mRNA AB002363.2 - 5 - 6 2630 85
AF518874.1 - 5 - 6 2467 85
AK074842.1 - 5 - 6 2403 85
AK291275.1 - 5 - 6 2445 85
AK299860.1 - 5 - 6 1893 85
AL832488.1 - 5 - 6 2398 85
BC020586.1 - 5 - 6 2398 85
BX647813.1 - 5 - 6 2407 85

View Full Product Details