The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000875.4 | NP_000866.1 | 21 - 21 | 11860 | 98 |
NM_001291858.1 | NP_001278787.1 | 21 - 21 | 11857 | 98 | |
XM_017022136.1 | XP_016877625.1 | 21 - 21 | 11363 | 98 | |
XM_017022137.1 | XP_016877626.1 | 22 - 22 | 10877 | 98 | |
XM_017022138.1 | XP_016877627.1 | 21 - 21 | 11360 | 98 | |
GenBank mRNA | AB425196.1 | - | 21 - 21 | 11341 | 98 |
BC010607.1 | - | 1 - 1 | 2871 | 98 | |
BC088377.1 | - | 1 - 1 | 891 | 98 | |
BC111046.1 | - | 1 - 1 | 1201 | 98 |