The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 13 - 14 | 1910 | 101 |
GenBank mRNA | AF021351.1 | - | 14 - 15 | 1777 | 101 |
AK026023.1 | - | 4 - 5 | 484 | 101 | |
AK300329.1 | - | 13 - 14 | 1890 | 101 | |
AY091460.1 | - | 6 - 7 | 685 | 101 | |
BC041089.1 | - | 13 - 14 | 1871 | 101 |