The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284301.2 | NP_001271230.1 | 12 - 13 | 2408 | 102 |
NM_001284302.2 | NP_001271231.1 | 12 - 13 | 2405 | 102 | |
NM_018685.4 | NP_061155.2 | 13 - 14 | 2519 | 102 | |
XM_006715746.1 | XP_006715809.1 | 13 - 14 | 2516 | 102 | |
XM_006715747.3 | XP_006715810.1 | 12 - 13 | 2405 | 102 | |
XM_017012354.1 | XP_016867843.1 | 13 - 14 | 2504 | 102 | |
XM_017012355.1 | XP_016867844.1 | 13 - 14 | 2504 | 102 | |
XM_017012356.1 | XP_016867845.1 | 12 - 13 | 2393 | 102 | |
GenBank mRNA | AF273437.1 | - | 13 - 14 | 2454 | 102 |
AK001468.1 | - | 3 - 4 | 438 | 102 | |
AK001472.1 | - | 8 - 9 | 1186 | 102 | |
AK023208.1 | - | 4 - 5 | 560 | 102 | |
AK291254.1 | - | 12 - 13 | 2337 | 102 | |
AK299803.1 | - | 13 - 14 | 2221 | 102 | |
BC034692.1 | - | 2 - 3 | 215 | 102 | |
BC070066.1 | - | 12 - 13 | 2390 | 102 | |
CR936650.1 | - | 10 - 11 | 1609 | 102 |