Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANLN
Entrez Gene ID
54443
Gene Name
anillin actin binding protein
Gene Aliases
FSGS8, Scraps, scra
Chromosome Location
Chr.7: 36389803 - 36453791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284301.2 NP_001271230.1 20 - 21 3086 134
NM_001284302.2 NP_001271231.1 20 - 21 3083 134
NM_018685.4 NP_061155.2 21 - 22 3197 134
XM_017012355.1 XP_016867844.1 21 - 22 3182 134
XM_017012356.1 XP_016867845.1 20 - 21 3071 134
GenBank mRNA AF273437.1 - 21 - 22 3132 134
AK001468.1 - 11 - 12 1116 134
AK001472.1 - 16 - 17 1864 134
AK023208.1 - 12 - 13 1238 134
AK291254.1 - 20 - 21 3015 134
AK299803.1 - 21 - 22 2899 134
BC034692.1 - 10 - 11 893 134
BC070066.1 - 20 - 21 3068 134
CR936650.1 - 18 - 19 2287 134

View Full Product Details