Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3RF3
Entrez Gene ID
344558
Gene Name
SH3 domain containing ring finger 3
Gene Aliases
POSH2, SH3MD4
Chromosome Location
Chr.2: 109129439 - 109504636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099289.2 NP_001092759.1 8 - 9 2146 63
XM_011511110.2 XP_011509412.1 6 - 7 1948 63
GenBank mRNA AK074131.1 - 6 - 7 1221 63
AL831825.1 - 1 - 2 134 63

View Full Product Details