Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TPI1P3
- Entrez Gene ID
-
728402
- Gene Name
- triosephosphate isomerase 1 pseudogene 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 116038731 - 116039944 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027338.1 |
|
1 - 1 |
558 |
149 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105377962 |
LOC105377962 |
|
-394.8kb
|
Anti-sense |
Chr. - 115633542 - 115643835 on Build GRCh38 |
Downstream |
5'-nucleotidase domain containing 1 |
NT5DC1 |
C6orf200, LP2642, NT5C2L1 |
60.9kb
|
Sense |
Chr. - 116100833 - 116245690 on Build GRCh38 |
View Full Product Details