Gene Transcripts

Gene Symbol
Pik3c2b
Entrez Gene ID
240752
Gene Name
phosphoinositide-3-kinase, class 2, beta polypeptide
Gene Aliases
C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta
Chromosome Location
Chr. 1: 133045672 - 133108688 on Build GRCm38
UniGene ID
Mm.387308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099276.2 NP_001092746.2 7, 8 2009
GenBank mRNA AK049181.1 - 7, 8 1982
BC132141.1 - 7, 8 1657 40130418

View Full Product Details