The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011075.2 | NP_035205.1 | 1 - 2 | 138 | 76 |
GenBank mRNA | AK030328.1 | - | 1 - 2 | 139 | 76 |
BC141363.1 | - | 1 - 2 | 95 | 76 | |
BC150811.1 | - | 1 - 2 | 95 | 76 | |
M14757.1 | - | 1 - 2 | 98 | 76 |