The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025407.2 | NP_079683.2 | 2 - 3 | 276 | 94 |
GenBank mRNA | AK002292.1 | - | 2 - 3 | 217 | 94 |
AK004031.1 | - | 2 - 3 | 222 | 94 | |
AK010553.1 | - | 2 - 3 | 216 | 94 | |
AK013128.1 | - | 2 - 3 | 216 | 94 | |
AK151764.1 | - | 2 - 3 | 218 | 94 | |
AK160337.1 | - | 2 - 3 | 223 | 94 | |
AK167911.1 | - | 2 - 3 | 216 | 94 | |
AK168218.1 | - | 2 - 3 | 218 | 94 | |
AK177346.1 | - | 1 - 2 | 69 | 94 | |
AK184915.1 | - | 1 - 2 | 69 | 94 | |
BB649589.1 | - | 2 - 3 | 276 | 94 | |
BC030064.1 | - | 2 - 3 | 213 | 94 | |
BY260300.1 | - | 2 - 3 | 249 | 94 |